Find methods for your needs
Forensic Screening for Drugs in Urine Using High-Resolution MS/MS Spectra and Simplified High-Performance Screening Software
Description
Forensic toxicologists need an economical solution to screen for a virtually unlimited number of compounds in urine. Here we present a method for screening using a dilute-and-shoot approach with the Q Exactive Focus mass spectrometer and Thermo Scientific ToxFinder software. For forensic use only.| Market: | Forensics; Toxicology |
| Keywords: | drugs of abuse, screening, Forensic Toxicology, Q Exactive Focus, ToxFinder |
| Matrix: | Urine |
| Author: | Marta Kozak, Kristine Van Natta |
| Affiliation: | Thermo Fisher Scientific |