Find methods for your needs
Determination of electrolytes in sports beverages by HPLC-CAD
Description
The Thermo Scientific Dionex UltiMate 3000 RS system is applied for the analysis of electrolytes in sports beverages. The separation is performed on a Thermo Scientific Acclaim Trinity P2 column, which is designed to resolve a broad range of anions and cations, mono- or multi-valent, in a single analysis using a simple gradient method. The Thermo Scientific OnGuard II P cartridge was used to remove artificial colors from the samples.| Market: | Food and Beverage |
| Keywords: | Acclaim Trinity P2, Acesulfame, Ascorbic acid, CAD, Calcium, Charged aerosol detection, Chloride, Citrate, Corona, Drink, Electrolytes, HPLC, Magnesium, OnGuard II P, Phosphate, Potassium, Sodium, Sports beverage, Sugars |
| Matrix: | Sports beverage |
For Research Use Only. Not for use in diagnostic procedures.