Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of the API (active pharmaceutical ingredient) amphetamine and its counterions. The separation is performed using a Thermo Scientific Acclaim Trinity P1 column, with both UV and charged aerosol detection. Amphetamine and saccharin respond to UV detection; aspartate, saccharin and sulfate respond to charged-aerosol detection. Note that metal contamination will cause peak tailing, and before use, the column should be cleaned with a chelating agent at pH 4.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, Amphetamine, API, Counterions, Saccharin, Aspartate, Charged aerosol detection, CAD, UV detection, Sulfate, Sodium, Adderall |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.