Find methods for your needs
Analysis of sugars in a sports beverage in HILIC mode using HPLC-CAD Be the first to rate this application
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of sugars in a sports beverage in HILIC mode. The separation is performed using a Thermo Scientific Acclaim Trinity P2 column, with charged aerosol detection. While ion analysis is its primary use, the Acclaim Trinity P2 can also be used in HILIC mode for analysis for hydrophilic neutral substances such as the simple sugars shown here.
Market: Food and Beverage
Keywords: Fructose, Glucose, HILIC, Charged aerosol detection, CAD, Maltose, Acclaim Trinity P2, Sodium, Sugars, Chloride
Matrix: Sports beverage
Uploaded on 5/8/2014.
For Research Use Only. Not for use in diagnostic procedures.