Find methods for your needs
Determination of High Sugar Concentrations in a Scotch Liqueur Sample Using a Compact Ion Chromatography System
Description
This record demonstrates the determinations of glucose, fructose, and sucrose in a 100-fold diluted liqueur alcoholic beverage sample by HPAE-PAD. Typically samples with g/L concentrations require >10,000-fold dilutions to remain in the linear range of the very sensitive HPAE-PAD technique. However, this method demonstrated on the Integrion RFIC system, uses a 0.4 µL internal injection loop with the High Concentration Carbohydrate Analysis Kit to extend the linearity from low mg/L to g/L concentrations. A wood hydrolysate application using this technique is attached for reference.| Market: | Food and Beverage |
| Keywords: | Carbohydrates, CarboPac PA20, Disaccharides, Glucose, Pulsed Amperometric Detection, HPIC, RFIC, Sucrose, PAD, fructose, monosaccharide, Alcoholic Beverages, HPAE-PAD, Integrion, Sugar, high performance anion-exchange chromatography, high concentration sugars, liqueur, Integrion HPIC, Integrion RFIC, Au on PTFE, distilled alcohol, ED detection, scotch, dextrose, TN21, TN 20, TN 70671, TN70671, AN1089, N20 |
| Matrix: | Alcoholic beverage |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.