Find methods for your needs
AN117: Quantification of Carbohydrates and Glycols in Pharmaceuticals - Small Sugars
Description
This application note describes the use of three different anion-exchange columns with amperometric detection to analyze common simple sugars, sugar alcohols, and glycols in pharmaceutical formulations. This record describes a method using the CarboPac PA10 column to determine some of the target carbohydrates. The reader is advised to review TN21 for updated waveform information.Market: | Pharma |
Keywords: | Cough Syrup, Glucose, CarboPac PA10, cold medicine, maltitol, pill, Sorbitol, tablet, Sucrose, mannitol, PAD, HPAE-PAD, TN21, TN71, TN110, TN 20, TN 70671, TN20, TN70671, TN 21 |
Matrix: | Pharmaceutical formulations |
Author: | Valoran Hanko and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.