Find methods for your needs
Quantitation of 122 Drugs in Urine by Triple Quadrupole Mass Spectrometry
Description
Forensic toxicologists face an ever-expanding list of compounds for analysis. This note presents work done using a next-generation triple quadrupole mass spectrometer with fast SRM acquisition speed for quantitation of 122 analytes in a single chromatographic run, where scan speed does not impact sensitivity or quantitative performance. Compounds analyzed include opiates, opioids, benzodiazepines, barbiturates, amphetamines, tricyclic antidepressants, illicit compounds, and more. For forensic use only.Market: | Forensics; Toxicology |
Keywords: | amphetamines, Barbiturates, benzodiazepines, drugs of abuse, opiates, Tricyclic antidepressants, TraceFinder, Forensic Toxicology, TSQ Endura, Opioids |
Matrix: | Urine |
Author: | Kristine Van Natta, Marta Kozak |
Affiliation: | Thermo Fisher Scientific |