Find methods for your needs
Trace Anion Analysis in Ultrapure Water Using an Electrolytic Water Purifier with a Compact Ion Chromatography System: Water Blank
Description
Determinations of trace anions in ultrapure water are important to the electronics industry. In this record, determinations of ng/L (ppt) anions using large volume concentration are made possible by the Electrolytic Water Purifier(EWP) and AutoPrep modules. Calibration standards are prepared using the AutoPrep large and small loops and EWP-purified water diluent, thereby increasing sensitivity by eliminating sources of contamination. The trace anions are separated on an IonPac AS17-C column designed to improve low level sulfate determinations using a Dionex Integrion compact IC system.| Market: | Other |
| Keywords: | Electrolytic water purifier, Formate, ppb, HPIC, AS-HV autosampler, Reagent-Free Ion Chromatography, fluoride, Nitrate, RFIC, trace anions, IC, Nitrite, Chloride, phosphate, IonPac AS17-C, Acetate, Sulfate, ultra pure water, suppressed conductivity detection, carbonate removal device, Reagent Free IC, carbonate removal, Carbonate, Integrion, Ion chromatograhy, AutoPrep, EWP, ppt, Integrion HPIC, Integrion RFIC, CRD 300, Ultrapure water |
| Matrix: | Ultra pure water, Water blank |
| Author: | Terri Christison and Daniel Khor |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.