Find methods for your needs
AN170: Determination of Silicate in High-Purity Water Using Ion Chromatography and Online Sample Preparation.
Description
The water used in the manufacture of semiconductors and other electronic components must be extremely pure, referred to as ultra high-purity water (UHPW). The production of UHPW includes deionization to remove corrosive strong acid anions. Deionization cartridges exhaust their capacity over time. To ensure the early recognition of cartridge depletion, silicate must be detected at concentrations lower than 1 μg/L. These low detections limits can be achieved by pre concentrating UHPW on a Dionex IonPac AG4A-SC column, then separating the concentrated sample on a Dionex IonPac AS17 column set.| Market: | Material Science |
| Keywords: | Cartridge Depletion, Concentrator column, IonPac AG4A-SC, IonPac AS17 C, Molybdate, Online sample preparation, PCR, Post column reaction, Preconcentration, Reagent Free Ion Chromatography, RFIC, Semiconductor Manufacturing, Silicates, UHPW, ultrahigh-purity water, Visible Absorbance |
| Matrix: | DI water |
For Research Use Only. Not for use in diagnostic procedures.