Find methods for your needs
A pre-concentration and on-line solid phase extraction setup for the LC-MS analysis of therapeutic protein mixtures
Description
This study shows a fully automated UHPLC setup applying a simple and efficient online solid phase extraction method. Showcase assay performance by using a column cartridge for sample trapping and a Thermo Scientific™ MAbPac™ RP column for separation with water/acetonitrile-based gradients. Leverage the new technologies such as the Thermo Scientific™ Vanquish™ UHPLC platform in combination with the Thermo Scientific™ Q Exactive™ HF Hybrid Quadrupole Orbitrap mass spectrometer.| Market: | BioPharma |
| Keywords: | Intact protein, Monoclonal antibody, Preconcentration, Vanquish, Biopharmaceutical, mass spectrometry, MAbPac, mAb, Q Exactive, Biomolecules, MSPac, Reversed phase, trap and elute, desalt |
| Matrix: | Drug product |
| Author: | Martin Samonig, Sabrina Patzelt, Martin Rühl, and Remco Swart |
| Affiliation: | Thermo Fisher Scientific, Germering, Germany |
For Research Use Only. Not for use in diagnostic procedures.