Find methods for your needs
Trace analysis of pharmaceuticals and organic contaminants in water
Description
We present the reliable and accurate quantitative analysis of contaminants at the pg/mL level in drinking water using the Thermo Scientific EQuan MAX Plus LC-MS system coupled to the Thermo Scientific TSQ Endura triple quadrupole mass spectrometer. Excellent reproducibility was shown for the target compounds in tap water using 1 mL injections at 0.37× maximum effluent concentration.| Market: | Water Analysis |
| Keywords: | Atenolol, Estrone, Ibuprofen, Linuron, Naproxen, Phenytoin, Trimethoprim, Water analysis, Drinking water, Environmental analysis, EQuan MAX Plus, Nonylphenol, TSQ Endura, Environmental Monitoring, PPCP |
| Matrix: | Water |
| Author: | Ken Matuszak, Kevin McHale, David Kage, Guoxin Lu |
| Affiliation: | Thermo Fisher Scientific; Water Quality Association |