Find methods for your needs
TN62: Reducing Carbonate Interference in Anion Determinations with the Carbonate Removal Device (CRD): 5 ppb Perchlorate in 500 mg/L Carbonate, Chloride, and Sulfate Matrix
Description
Carbonate can interfere with accurate quantification of adjoining anions, such as sulfate and perchlorate. Perchlorate is a highly regulated contaminant (EPA Method 314) because of the health risks to pregnant women and prenatal and developing children. Here we show the efficient removal of the carbonate peak from a synthetic high ionic water sample using the Dionex CRD 200 device. In this example, the high ionic water matrix contains: 5 ppb perchlorate in 500 mg/L each of carbonate, sulfate and chloride.| Market: | Environmental |
| Keywords: | anions, carbonate, carbonate removal device, chloride, CRD 200 device, high ionic water, HIW, IC, ion chromatograhy, IonPac AS16 column, perchlorate, Reagent Free IC, RFIC, sulfate, EPA Method 314 |
| Matrix: | 500 mg/L carbonate, Chloride, Sulfate matrix (high ionic water) |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.