Find methods for your needs
API and counterions in Adderall using multi-mode liquid chromatography with charged aerosol detection
Description
This work demonstrates the determination of the API (amphetamine) and counterions (aspartate, saccharin and sulfate) in a medicine Adderall within a single analysis using a Thermo Scientific Acclaim Trinity P2 column and Thermo Scientific Dionex Corona Veo Charged Aerosol Detector (CAD).| Market: | Pharma |
| Keywords: | Counterion, Acclaim Trinity P2, Adderall, Amphetamine, Charged aerosol detection, HPLC |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.