Find methods for your needs
AN116: Quantification of Anions in Pharmaceuticals (Hydroxide Eluent) Be the first to rate this application
an116 quantification anions pharmaceuticals hydroxide eluent
No compound information available.
Description
The United States Food and Drug Administration (U.S. FDA) and regulatory agencies in other countries require that pharmaceutical products be tested for composition. This Application Note describes the use of two anion exchange columns with suppressed conductivity detection to analyze common anions in pharmaceutical formulations. Here demonstrate that the IonPac AS11 resolves, in the same injection, common inorganic anions such as fluoride, chloride, bromide, sulfate, nitrate, and phosphate, as well as common organic anions such as benzoate, sorbate, citrate, and saccharin.
Market: Pharma
Keywords: Anions, benzoate, bromide, chloride, citrate, FDA, fluoride, Ion chromatography, nitrate, pharmaceutical, phosphate, saccharin, sorbate, sulfate, L61
Matrix: Drug
Uploaded on 6/23/2015.
For Research Use Only. Not for use in diagnostic procedures.