Find methods for your needs
AN1107: Determination of Anions and Carboxylic Acids in Urban Fine Particles (PM2.5)
Description
Ionic forms of air pollutants are analyzed here using anion-exchange chromatography and suppressed conductivity. The gas sample included particles < 2.5 um (PM2.5) was collected using ambient ion monitoring and then dissolved in deionized water. The dissolved inorganic anions and organic acids were separated on a 4 mm i.d. IonPac AS11-HC column and detected by suppressed conductivity. This method provided analysis of more ionic contaminants than standard methods (EPA Method 26A) and ASTM D5085-02). An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | Carboxylic acid, Formate, ICS-5000+, Organic acids, HPIC, fluoride, air analysis, air sampling, mesylate, inorganic anions, ambient air, air particles, Succinate, Malonate, Nitrate, RFIC, PM2.5, IC, Chloride, Oxalate, Acetate, IonPac AS11-HC, Sulfate, ICS-6000 |
| Matrix: | Aerated water |
| Author: | Liao Dongli, Xu Qun, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.