Find methods for your needs
AN185: Determination of Trace Organic Acids and Inorganic Anions in Boric Acid-Treated Power Plant Waters Using an Automated Reagent-Free Ion Chromatography System
Description
Boron as boric acid is used to control the nuclear reaction in pressurized water reactors because of its strong neutron-absorbing characteristics. To prevent corrosion, anionic contamination is monitored. Here sub-ppb concentrations of inorganic anions in a large volume sample are determined in borated waters using matrix elimination through the use of a CR-CTC trap column. The anions were collected on a concentrator column, eluted and determined using an electrolytically generated hydroxide gradient on IonPac AS15 column. This is an improvement over using borated eluents (AN166).| Market: | Water Analysis |
| Keywords: | Acetate, Chloride, Glycolate, sulfate, fluoride, anions, boric acid, boron, lithium, organic acids, power plant, RFIC, Trace analysis, Nitrite, Phosphate, Bromide, Nitrate, PWR, borate |
| Matrix: | Borated power plant water |
For Research Use Only. Not for use in diagnostic procedures.