Find methods for your needs
TN122: Separation of Heat Stable Amine Salts in Methyldiethanolamine (MDEA) SolutionsUsing High-Pressure IC
Description
Sour gas is treated with amine and recycled amine (scrubbing) solutions to absorb toxic and undesirable H2S and CO2 create sweet natural gas. Salts of organic acids and sulfur species, heat stable amine salts (HSAS) can concentrate over time. Analysis of these HSAS are needed to assess the efficiency of scrubbing solution and can be quite challenging. In this technical note, HSAS in methyldiethanolamine (MDEA) were separated with high efficiency on a 2 mm i.d. IonPac AS11HC-4μm column using a high pressure ion chromatography system.| Market: | Petrochem, Oil and Gas |
| Keywords: | Bromide, Chloride, Formate, Formate, Nitrate, Nitrite, Oxalate, Phosphate, Propionate, Sulfate, Thiocyanate, Thiosulfate, HSAS, Fluoride, heat stable salts, heat stable amine salts, anions in amine solutions, sour gas, IonPac AS11-HC-4um column, IonPac AS11-HC column, Sulfite |
| Matrix: | Methyldiethanolamine |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.