Find methods for your needs
Simple gradient method for the analysis of calcium and magnesium in a dietary mineral supplement using HPLC-CAD
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of calcium and magnesium in a dietary mineral supplement. The separation is performed using a Thermo Scientific Acclaim Trinity P2 column, with charged aerosol detection. The Thermo Scientific Acclaim Trinity P2 column offers ideal selectivity for various cations and anions including organic acids. In this case, all label claimed minerals and their counterions are confirmed.| Market: | Food and Beverage |
| Keywords: | Counterions, Magnesium, Aspartate, Citrate, Charged aerosol detection, CAD, Calcium, Acclaim Trinity P2 |
| Matrix: | Mineral supplement tablet |
For Research Use Only. Not for use in diagnostic procedures.