Find methods for your needs
TN72206: Configuring the Dionex Modular HPIC System with an Inline Water Purifier and Inline Calibration for Trace Anion Determinations in Ultrapure Water: EWP Water Blank
Description
In the electronics industry, ionic contamination in the range of parts per trillion (ppt, ng/L) to parts per billion (ppb) concentrations is a major concern, causing corrosion-related failures in production and final product. Ionic contamination is increasingly important as the devices decrease to size. This technical note demonstrates automated calibration and water purification on an ICS-5000+ HPIC dual IC system to minimize the system and environmental contamination which resulted in increased sensitivity to double-digit-ppt concentrations. An ICS-6000 can be used for this application.| Market: | Chemical |
| Keywords: | Electrolytic water purifier, Formate, Reagent Free Ion Chromatography, Reagent Free Ion Chromatography (RFIC), HPIC, ICS-5000+ HPIC system, AS-HV autosampler, fluoride, RFIC, trace anions, IC, Chloride, Sulfate, Reagent Free IC, AutoPrep, EWP, ICS-5000+ IC system, RFIC System, RFC 10, IonPac AS17-C column, IonPac AS17C column, ICS-6000, TN 73892, TN73892, TN177 |
| Matrix: | Deionized water purified inline by EWP |
| Author: | Terri Christison, Daniel Khor, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |