Find methods for your needs
AN1112: Determination of Anions in Breast Milk
Description
Breast milk is the primary source of nutrition for newborns before they are able to eat and digest other foods, so there is a close correlation between breast milk quality and infant health. In addition to proteins and vitamins, the ionic elements such as sodium, potassium, calcium, magnesium, chloride, and nitrate in breast milk also are important for infant health. However, consumption of breast milk that contains a high concentration of these ions could lead to health issues. IC is an efficient means to determine anions. An ICS-6000 can be used for this application.| Market: | Clinical Research |
| Keywords: | ESI, ICS-5000+, MSQ, Citrate, Nitrate, RFIC, SIM, IC-MS, IonPac AS19, Chloride, phosphate, ICS-6000 |
| Matrix: | Breast milk |
| Author: | Liao Dongli, Xu Qun, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |