Find methods for your needs
Separation of Vitamin C, Vitamin B12, Folic Acid and Vitamin-E from Multivitamin Capsules.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation of multivitamins (Folic Acid,Vitamins C, B12, and E from a Multivitamin Capsule. The separation was performed on a Thermo Scientific Acclaim Polar Advantage-II HPLC column with a UV detection at 270 nm. With a run time of 35 minutes, the method allows easy separation of Vitamin C, B12, Folic Acid and E from Multivitamin Capsule.| Market: | Pharma |
| Keywords: | Capsule, Folic acid, HPLC, UV detection, Vitamin-B12, Multivitamins, Vitamin-C, Vitamin-E, Acclaim Polar Advantage-II |
| Matrix: | Formulation |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.