Find methods for your needs
AN1149: Profiling Fructosyloligosaccharides (FOS)-containing samples by HPAE-PAD
Description
In this work, the oligo- and polysaccharide distribution of three different inulin samples has been characterized by HPAE-PAD. In addition, HPAE-PAD profiling was demonstrated as a method to determine FOS content in prebiotic samples and variation in prebiotics on addition of probiotics. We used Inulin-FOS and chicory root inulin as prebiotic samples. For probiotics, we used Yakult drink, a well known probiotic dairy product. In this method, separation of individual oligosaccharides in FOS was achieved on a Dionex CarboPac PA200 column. An ICS-6000 can be used for this application.| Market: | Food and Beverage |
| Keywords: | Dietary supplements, ICS-5000+, inulin, oligosaccharide, HPAEC-PAD, Dionex CarboPac PA200 column, prebiotics, probiotics, Disposable electrode, Fructooligosaccharide (FOS), Yakult drink, Lactobacillus casei Shirota, Chicory root inulin, Dahlia Tuber inulin, ICS-6000, TN 20, TN 70671, TN20, TN70671, AN 1149 |
| Matrix: | Food supplements |
| Author: | Manali Aggrawal and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.