Find methods for your needs
Determination of Inorganic Anions in Environmental Waters Using a Compact Ion Chromatography System
Description
This method shows the determination of inorganic anions in an environmental water sample, previously published in Application Note 154. In this record, the method is executed on an Integrion HPIC system with eluent generation.| Market: | Environmental |
| Keywords: | HPIC, waste water, fluoride, anions, Nitrate, RFIC, IC, Nitrite, Chloride, phosphate, Sulfate, Suppressed Conductivity, EPA 300.1, wastewater, IonPac AS18 column, Integrion, Ion chromatograhy, municipal drinking water, Integrion HPIC, Integrion RFIC |
| Matrix: | Municipal drinking water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.