Find methods for your needs
Determination of Sulfate and Sulfamate in Topiramate Using a Compact Reagent-Free Ion Chromatography System
Description
Determination of sulfamate and sulfate are used to monitor and confirm topiramate degradation. In application note 238 we demonstrate the use of a Reagent-Free Ion Chromatography (IC) system with a Thermo Scientific Dionex IonPac AS11 column and suppressed conductivity detection for the determination of sulfamate and sulfate in topiramate. This record shows that the method can be run using a new compact IC system: the Thermo Scientific Dionex Integrion system.| Market: | Pharma |
| Keywords: | Pharmaceutical, Supressed conductivity detection., degradation, Impurities, Dionex IonPac AS11 column, RFIC, Drug, Intergrion, application note 238, Topiramate |
| Matrix: | Drug |
| Author: | Hua Yang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.