Find methods for your needs
AB135: Determination of Anions and Organic Acids in Brewed Coffee Samples Using Capillary IC
Description
The characteristic aroma, acidity, and flavor of a coffee are attributed to the inorganic anions, organic acids, chlorogenic acid, and monosaccharides content. In this study, inorganic anions and organic acid anions in caffeinated and decaffeinated brewed coffee samples are separated within 20 min by anion-exchange chromatography on a capillary Dionex IonSwift MAX-100 (0.25 × 250 mm) column set using a gradient of potassium hydroxide eluent at 12 μL/min.Market: | Food and Beverage |
Keywords: | Coffee, Formate, Fumarate, Glutarate, Malate, Propionate, Quinate, Citrate, Isocitrate, Nitrate, RFIC, Chloride, Lactate, Oxalate, phosphate, Acetate, Sulfate, IonSwift MAX-100, Bromide, Itaconate, caffeinated, decaffeinated, Arabica, Robusta |
Matrix: | Brewed coffee |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.