Find methods for your needs
TN62: Reducing Carbonate Interference in Anion Determinations with the Carbonate Removal Device (CRD): Trace Anions
Description
Carbonate can interfere with accurate quantification of adjoining anions, such as sulfate. Here we show the efficient removal of the carbonate peak from a trace analysis water sample using the Dionex CRD 200 device. In this example, a large volume (1000 uL) of water spiked with < 4 ug/L concentration of 17 anions is directly analyzed.| Market: | Environmental |
| Keywords: | Phthalate, Glycolate, adipate, Bromide, acetate, anions, carbonate, carbonate removal device, CRD 200 device, fluoride, formate, IC, ion chromatography, IonPac AS15-5 µm column, nitrate, oxalate, phosphate, Reagent-Free IC, RFIC, sulfate, trace anion analysis, methanesulfonate, HIBA, hydroxyisobutyrate |
| Matrix: | Pure water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.