Find methods for your needs
TN62: Reducing Carbonate Interference in Anion Determinations with the Carbonate Removal Device (CRD): Perchlorate in Drinking Water
Description
Carbonate can interfere with accurate quantification of adjoining anions, such as perchlorate and sulfate. Perchlorate is a highly regulated contaminate because of the health risks to pregnant women, prenatal and developing children. Here we show the efficient removal of the carbonate peak from a synthetic high ionic water sample using the Dionex CRD 200 device. In this example, the high ionic water matrix contains: 5 ppb perchlorate in a Sunnyvale drinking water sample.| Market: | Environmental |
| Keywords: | Chloride, EPA Method 314.0, Sulfate, anions, carbonate, carbonate removal device, CRD 200, drinking water, IC, ion chromatography, IonPac AS16 column, perchlorate, RFIC, suppressed conductivity |
| Matrix: | 5 ppb perchlorate in drinking water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.