Find methods for your needs
AN92: The Determination of Sugars in Molasses by High-Performance Anion Exchange with Pulsed Amperometric Detection
Description
The accurate measurement of the amount of sugar in final molasses is economically important to sugar mills, because the purchase price of the final molasses is directly related to its sugar content. A high-performance anion exchange method with pulsed amperometric detection (HPAE-PAD) was developed to accurately determine the sugar concentrations of cane and beet final molasses samples.| Market: | Food and Beverage |
| Keywords: | AOAC, carbohydrate, CarboPac PA1, Electrochemical detection, food, Glucose, Raffinose, Sucrose, PAD, cane sugar, ICUMSA, cane molasses, beet sugar, beet molasses, fructose, HPAE-PAD, TN21, TN 20, TN 70671, TN20, TN70671, TN110:TN71 |
| Matrix: | Sugar |
| Author: | Jill Jekot |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.