Find methods for your needs
Direct Determination of Unlabelled Carbohydrates, used in Biotech Formulations by HPLC-CAD
Description
In many of the vaccines and other formulations that are used in the prevention and treatment of many conditions and illnesses contain carbohydrates, as a part of their formulation. Six carbohydrates were identified as important components in biotechnological formulations: sorbitol, mannitol, sucrose, lactose, maltose, and trehalose. These are separated by HILIC-HPLC and detected by the universal Corona charged aerosol detector.| Market: | BioPharma |
| Keywords: | Carbohydrates, Maltose, Sorbitol, Lactose, Sucrose, mannitol, trehalose |
| Matrix: | Water |
| Author: | M Plante |
For Research Use Only. Not for use in diagnostic procedures.