Find methods for your needs
AN2967: Implementation of a Walk-Up High-Pressure Capillary Ion Chromatograph for the Fast Separation of Pharmaceutical Relevant Inorganic Cations (pain relief tablet)
Description
The counter ion impacts the API drug’s efficacy, solubility, and toxicity, and therefore counter ion determinations are important to pharmaceutical companies during all stages of product development and as part of product quality monitoring. In this example, the sodium counter ion is determined in a water extract of a pharmaceutical drug tablet designed for pain relief. Here fast sodium determinations were demonstrated on an IonPac CS12A-8µm capillary column using a walk-up high pressure capillary IC system.| Market: | Pharma |
| Keywords: | IonPac CS12A column, Sodium, cap IC, capillary IC, counter cations, counter ion, fast IC, IonPac CS12A-8um column, NSAID |
| Matrix: | Water extract of pharmaceutical drug tablet developed to relieve pain |
| Author: | T. Christison, J.E. Madden, F. Pang, and K. Divan |
For Research Use Only. Not for use in diagnostic procedures.