Find methods for your needs
AN247 (a): Determination of Morpholine, Ethanolamine, and Hydrazine in Simulated Nuclear Power Plant Wastewater
Description
This Application Note describes the determination of μg/L concentrations of hydrazine, morpholine, and ethanolamine (ETA) in a simulated NPP wastewater sample containing mg/L concentrations of common cations. Morpholine and hydrazine are separated using an IonPac CS16 column with electrolytically generated methanesulfonic acid eluent. See AN 247 (b) for ETA separation on an IonPac CS15 column.| Market: | Water Analysis |
| Keywords: | Amines, ammonium, Electrochemical detection, IonPac CS16, Magnesium, Nuclear Power Plant, PCR, Post column reaction, Potassium, Pulsed Amperometric Detection, Hydrazine, PAD, morpholine, Calcium, Lithium, Sodium, ETA, Corrosion Inhibitors, oxygen scavenger |
| Matrix: | Wastewater |
| Author: | Terri Christison, Brian DeBorba, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.