Find methods for your needs
AN106: Ion (Ion Exclusion) Chromatography in the Pharmaceutical Industry
Description
Ion Chromatography is a simple and accurate technique for the analysis and quantification of charged species. This application note describes the use of ion exchange chromatography and ion exclusion chromatography in the pharmaceutical industry for the analysis of ions in ingredients and final products. The topic of cleaning validation is also discussed as it applies to charged species. Here shows the separation of several weak organic acids by ion exclusion chromatography using an IonPac ICE-AS6 column.| Market: | Pharma |
| Keywords: | Acetate, a-Hydroxyisobutyric acid, Anions, Citrate, Formate, Fumarate, IC, Ion exclusion, IonPac ICE-AS6 column, Lactate, Malate, Oxalate, Proprionate, Succinate, Tartrate |
| Matrix: | Drug |
For Research Use Only. Not for use in diagnostic procedures.