Find methods for your needs
AN61: Determination of Tobramycin and Impurities Using HPAE-PAD
Description
In this application note, we combine the CarboPac PA1, eluent generator with CR-ATC, and disposable Au working electrodes to demonstrate an improved HPAE-PAD technology for tobramycin purity analysis.| Market: | Pharma |
| Keywords: | aminoglycoside, aminoglycoside antibiotic, Antibiotic, carbohydrate, CarboPac PA1, Drug Substance, kanamycin, kanamycin B, nebramine, neamine, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 61 |
| Matrix: | Drug |
| Author: | Valoran Hanko and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.