Find methods for your needs
AN280: Carbohydrate in Coffee: A New Fast HPAE-PAD Method
Description
A fast method using the Dionex CarboPac SA10 column was tested for determining the common coffee carbohydrates. This column achieves fast, high resolution separation of mono- and disaccharides commonly found in food samples. The Dionex CarboPac SA10 column is composed of a wide-pore macroporous substrate coated with a strong anion-exchange latex of nano-beads. The combination of the high capacity provided by the substrate and the new internal chemistry of the nano-bead functionality delivers high resolution and short analysis time for the common sugars of interest in food and beverages.| Market: | Food and Beverage |
| Keywords: | AOAC, arabinose, carbohydrate, Carbohydrates, CarboPac PA1, Coffee beans, disaccharide, EG, Eluent Generation, fucose, Glucose, green coffee, instant coffee, rhamnose, ribose, xylose, Mannose, Sucrose, mannitol, PAD, fructose, monosaccharide, galactose, HPAE-PAD, AN 280, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Coffee |
| Author: | Lipika Basumallick and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.