Description
The Thermo Scientific Acclaim Trinity P2 provides HILIC interactions in addition to anion-exchange and cation-exchange properties. It is shown here that both monosaccharides (fructose and glucose) and disaccharides (sucrose and lactose) can be sufficiently retained and separated on a 50 mm long column in 80% acetonitrile at 60 °C.| Market: | Food and Beverage |
| Keywords: | Disaccharides, Fructose, Glucose, HPLC, Monosaccharides, Charged aerosol detection, Lactose, Sucrose, Acclaim Trinity P2 |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.