Description
The Thermo Scientific Acclaim Trinity P2 column was used to determine a small amount of guanidine in a background of phosphate buffered saline (PBS). Even though the high concentration of sodium chloride overloads the column, it has sufficient resolution for quantifying all the components.| Market: | BioPharma |
| Keywords: | HPLC, Charged aerosol detection, Acclaim Trinity P2, Phosphate buffered saline (PBS), Guanidine |
| Matrix: | Phosphate buffered saline (PBS) |
For Research Use Only. Not for use in diagnostic procedures.