Find methods for your needs
Increased resolution of tryptic digests of proteins using a Thermo Scientific Acclaim RSLC C18 HPLC column
Description
In this work, a digestion of bovine serum albumin is resolved using the Thermo Scientific Acclaim RSLC C18 into 127 peaks with an average peak width of 0.27 minutes. For a 90 minute gradient, this works out to a theoretical peak capacity of >300.| Market: | BioPharma |
| Keywords: | Acclaim RSLC C18, Bovine serum albumin, HPLC, Ovalbumin, Protein, Tryptic digests, UV detection |
| Matrix: | Protein digest in TRIS buffer |
For Research Use Only. Not for use in diagnostic procedures.