Find methods for your needs
Assay for water-soluble vitamins in vitamin tablets using a polar-embedded HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of water-soluble vitamins in vitamin tablets. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with UV detection at various wavelengths. Some vitamins are strongly hydrophilic, so the column needs to operate in 100% aqueous buffer to gain sufficient retention to resolve them from the matrix. The Acclaim PA can do this reliably whereas a hydrophobic C18 column would likely suffer dewetting.
Market: Food and Beverage
Keywords: Acclaim PolarAdvantage (PA), Ascorbic acid, Folic acid, HPLC, Niacinamide, Pantothenic acid, Pyridoxine, Riboflavin, Thiamine, UV detection, Vitamin B12, Water-soluble vitamins
Matrix: Tablet (vitamin)
Uploaded on 2/10/2010.
For Research Use Only. Not for use in diagnostic procedures.