Find methods for your needs
Separation of oxidized and deamidated impurities of Bevacizumab using Hydrophobic Interaction Chromatography (HIC).
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Hydrophobic Interaction (HIC) analysis of Bevacizumab. The separation was performed on a Thermo Scientific MabPac HIC-20 column at UV detection of 214 nm. With a total run time of 40 minutes the method allows separation of all oxidized and deamidated impurities of Bevacizumab.| Market: | BioPharma |
| Keywords: | HPLC, Bevacizumab, PDA detection, Oxidation, Deamidation, MabPac HIC-20 |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.