Find methods for your needs
Separation of Oxidized and Deamidated Impurities from Rituximab by using Hydrophobic Interaction Chromatography (HIC)
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Hydrophobic Interaction (HIC) analysis of Rituximab. The separation was performed on a Thermo Scientific MabPac HIC-20 column at UV detection of 214 nm. With a total run time of 38 minutes the method allows separation of all oxidized and deamidated impurities of Rituximab.| Market: | BioPharma |
| Keywords: | HPLC, Hydrogen peroxide, PDA detection, Rituximab, Oxidation, Deamidation, MabPac HIC-20 |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.