Find methods for your needs
Determination of Carbohydrates in Coffee Using a New Fast HPAE-PAD Method
Description
This record demonstrates a fast, high resolution HPAE-PAD method based on the method in AN280 for determining common sugars of interest in food and beverages. The IC instrument described in AN280, is updated to the Thermo Scientific Dionex Integrion HPIC system. The CarboPac SA10 column used in Application Note 280 is replaced with CarboPac SA10-4μm column which delivers faster and higher resolution separations. This method was applied to coffer samples.| Market: | Food and Beverage |
| Keywords: | carbohydrate, Electrochemical detection, RFIC, HPAE-PAD, Carbopac SA10-4μm, Coffee Sugars, AN280, AOAC Method 995.13, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Water |
| Author: | Sachin Patil and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.