Find methods for your needs
CAN109: Primary Metabolite Analysis of Plant Material Using a Triple Quadrupole MS Coupled to a Monolith Anion-Exchange Column
Description
A platform using Dionex ion chromatography (IC) coupled to triple quadrupole mass spectrometry (MS) was developed to measure various metabolites in Brassica napus seeds. The IC tandem mass spectrometry (IC-MS/MS) technique provides a good tool to identify and quantify metabolites in a complex matrix.| Market: | Metabolomics |
| Keywords: | AMP, Fumarate, Trans-aconitate, Citrate, Succinate, Isocitrate, IonPac AS11, NADPH, 6-phosphogluconate, 2-oxoglutarate, CAN109, Cis-aconitrate, IonSwift MAX-100, UDP |
| Matrix: | Water |
| Author: | Nicolas Heinzel and Hardy Rolletschek |
| Affiliation: | Leibniz Institute of Plant Genetics and Crop Plant Research (IPK), Gatersleben, Germany |
For Research Use Only. Not for use in diagnostic procedures.