Find methods for your needs
TN55: Screening of Sample Matrices and Individual Matrix Ingredients for Suitability in AAA-Direct
Description
In this technical note, we describe a procedure to screen sample matrices for their effect on peak area, peak height, and retention time. We demonstrate this procedure by testing ethanol for possible interferences in AAA-Direct. However, the method described here can also be used to evaluate any matrix component analyzed by chromatography with electrochemical detection. In this technical note, we use the term test sample (TS) to describe the specific test compound evaluated for possible interferences.| Market: | BioPharma; Proteomics; Clinical Research |
| Keywords: | Antibiotics, Carbohydrates, IPAD, AminoPac PA10, TN55 |
| Matrix: | DI water |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.