Find methods for your needs
TN125: Guidelines for Successful Use of Thermo Scientific Dionex AminoTrap Columns
Description
Free amino acids and small peptides can interfere with HPAE-PAD monosaccharide analysis. An AminoTrap column in front of a CarboPac column can remove the aforementioned interference. This TN describes how to use the AminoTrap and how to test if it functioning correctly. The record reports an AminoTrap test method.| Market: | BioPharma |
| Keywords: | AminoTrap, carbohydrate, CarboPac, Electrochemical detection, glycoprotein, PAD, gold working electrode, monosaccharide analysis, HPAE-PAD, TN71, TN 71, TN 125, TN 20, TN 70671, TN20, TN70671, TN125 |
| Matrix: | Glycoprotein |
| Author: | Lipika Basumallick, Uday Aich, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.