Find methods for your needs
AU197: Anion Determinations in Municipal Wastewater Samples Using EPA Method 300.1 (A) on an Integrated Ion Chromatography System
Description
Inorganic anions were determined in three municipal wastewater samples according to U.S. EPA Method 300.1 (A). Nine anions were separated on IonPac AS22 anion-exchange column using manually prepared carbonate-bicarbonate eluent. The anions were detected by suppressed conductivity using chemical regenerant mode with the Thermo Scientific Dionex MMS 300 suppressor and 25 mN sulfuric acid at 7 mL/min.| Market: | Environmental |
| Keywords: | acetate, bromide, carbonate, carbonate eluent, chloride, fluoride, formate, IC, ion chromatography, IonPac AS22 column, nitrate, nitrite, phosphate, sulfate, suppressed conductivity detection, wastewater, PO4-P, NO3-N |
| Matrix: | Municipal wastewater samples |
| Author: | T.Christison, C.Tanner, J.E. Madden, L. Lopez |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.