Find methods for your needs
TN113: Practical Guidance for Using Capillary Anion Chromatography - Concentrate Mode
Description
Configuring a capillary IC system for trace analysis is discussed in this technical note, TN113. Tips and tricks for trace anion analysis for ion chromatography (IC) systems and capillary IC systems are discussed. Data is also shown for concentrating and direct injection of a large volume injection. This example demonstrates anion determinations by concentrating 200 uL of sample.| Market: | Water Analysis |
| Keywords: | acetate, bromide, capillary, capillary IC, chloride, fluoride, formate, IC, ICW-3000 Inline Purifier, Ion Chromatography, IonPac AS15 column, IonPac MAC 100 concentrator column, nitrate, nitrite, oxalate, phosphate, sulfate, trace analysis, trace anions, monolith column |
| Matrix: | Pure water |
| Author: | Christison, Schibler, Madden, Shao, Pang, Lopez |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.