Find methods for your needs
Fast, high resolution size exclusion separation of a protein mix by bio-compatible UHPLC
Description
A mix of five proteins in varying size was separated on a Thermo Scientific MabPac SEC-1 column using a Thermo Scientific Vanquish UHPLC system. The molecular weight range was 0.13 kDa – 670 kDa. Elution with high resolution of all five proteins was achieved within 15 minutes.| Market: | BioPharma |
| Keywords: | Ovalbumin, Protein, Size exclusion, UV detection, 4-Aminobenzoic acid (PABA), Ribonuclease, UHPLC, Thyroglobulin, gamma-globulin |
| Matrix: | N/A |
| Author: | Valeria Barattini |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.