Find methods for your needs
AN2967: Implementation of a Walk-Up High-Pressure Capillary Ion Chromatograph for the Fast Separation of Pharmaceutical Relevant Inorganic Anions (diabetes tablet)
Description
The counter ion impacts the API drug’s efficacy, solubility, and toxicity, and therefore counter ion determinations are important to pharmaceutical companies during all stages of product development and as part of product quality monitoring. In this example, the chloride counter ion is determined in a water extract of a pharmaceutical drug tablet designed to treat Type II diabetes patients. Here fast chloride determinations were demonstrated on an IonPac AS19 capillary column using a walk-up high pressure capillary IC system.Market: | Pharma |
Keywords: | cap IC, capillary IC, chloride, counter anion, counter ion, diabetes, fast IC, IonPac AS19 column, tablet |
Matrix: | Water extract of a pharmaceutical drug tablet to treat Type II diabetes |
Author: | T. Christison, J.E. Madden, F. Pang, and K. Divan |
For Research Use Only. Not for use in diagnostic procedures.