Find methods for your needs
AN83: Size-Exclusion Chromatography of Polysaccharides with Pulsed Amperometric Detection (PAD).
Description
Size-exclusion chromatography is used in carbohydrate research to determine molecular weight distributions of polysaccharides. Size-exclusion columns separate biological macromolecules by hydrodynamic size. Pulsed amperometric detection (PAD), provides sensitive and specific detection of carbohydrates. Here, fractionated pullulan standards were used to determine the separation range of linear carbohydrates.This application note shows how SEC can be coupled to PAD.| Market: | BioPharma |
| Keywords: | Carbohydrates, Fructose, Galactose, Glucose, inulin, Polysaccharides, Postcolumn reagent, Pulsed Amperometric Detection, Size-Exclusion Chromatography, Mannose, Gum arabic, Maltotriose, PAD, Agar, gum ghatti, starch, Pullulan |
| Matrix: | Gum ghatti, Locust bean gum |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.