Description
The separation of five sugars was performed using the Thermo Scientific Hypersil GOLD Amino column in HILIC mode with RI detection.| Market: | Food and Beverage |
| Keywords: | Fructose, Glucose, HPLC, Hypersil GOLD Amino, Lactose, Maltose, RI detection, Sucrose, Sugars |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.